Human recombinant SEMA3A (from cells)
Fournisseur: ProSci Inc.
91-936
PRSI91-936EA 584 EUR InStock
PRSI91-936
Human recombinant SEMA3A (from cells)
Protéines et Peptides
emaphorin-3A is a secreted protein which belongs to the semaphorin family. Semaphorins are a family of secreted and cell-bound signaling molecules defined by the presence of a common 500 aa Sema domain.This protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité