Human recombinant tPA (from cells)
Fournisseur: ProSci Inc.
91-938
PRSI91-938EA 852 EUR InStock
PRSI91-938
Human recombinant tPA (from cells)
Protéines et Peptides
Tissue-type plasminogen activator (PLAT) is a protein that secreted into extracellular space. PLAT contains five domains: EGF-like domain, fibronectin type-I domain, 2 kringle domains and peptidase S1 domain. It belongs to the peptidase S1 family. The main function of this protein is to convert plasminogen into biologically active plasmin. As a protease, PLAT plays a crucial role in regulating blood fibrinolysis, maintaining the homeostasis of extracellular matrix and in modulating the post-translational activation of growth factors. PLAT is found not only in the blood, where its primary function is as a thrombolytic enzyme, but also in the central nervous system (CNS). It participates in a number of physiological and pathological events in the CNS, as well as the role of neuroserpin as the natural regulator of PLAT's activity in these processes. Increased or decreased activity of PLAT leads to hyperfibrinolysis or hypofibrinolysis, respectively. In addition, as a cytokine, PLAT plays a pivotal role in the pathogenesis of renal interstitial fibrosis through diverse mechanisms. Thus, as a fibrogenic cytokine, it promotes the progression of kidney diseases.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité