Human recombinant VSIG4 (from cells)
Fournisseur: ProSci Inc.
91-940
PRSI91-940EA 528 EUR InStock
PRSI91-940
Human recombinant VSIG4 (from cells)
Protéines et Peptides
V-set and immunoglobulin domain-containing protein 4(VSIG4) is a transmembrane protein contains a signal peptide, a V-type Ig-like domain, a C2-type Ig-like domain, several potential O-glycosylation sites, and an intracellular domain with 2 potential phosphorylation sites and is structurally related to the B7 family of immune regulatory proteins. This protein is also a receptor for the complement component 3 fragments C3b and iC3b.The main function is strong negative regulator of T-cell proliferation and IL2 production and it is also potent inhibitor of the alternative complement pathway convertases. It abundantly expressed in several fetal tissues such as adult tissues, highest expression in lung and placenta and it also expressed in resting macrophages.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité