Mouse recombinant Ephrin B2
Fournisseur: ProSci Inc.
91-941
PRSI91-941EA 306 EUR InStock
PRSI91-941
Mouse recombinant Ephrin B2
Protéines et Peptides
Ephrin-B2 is a single-pass type I membrane protein and it contains 1 ephrin RBD (ephrin receptor-binding) domain. Ephrin-B2 belongs to the ephrin (EPH) family and it is cell surface transmembrane ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. The ephrins and EPH-related receptors contain the largest subfamily of receptor protein-tyrosine kinases and have been associated with mediating developmental events, particularly in the nervous system and in erythropoiesis. Based upon their structures and sequence relationships, ephrins are allocated into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. It also binds to receptor tyrosine kinase including EPHA4, EPHA3 and EPHB4 and together with EPHB4 plays a central role in heart morphogenesis and angiogenesis through regulation of cell adhesion and cell migration.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité