Human recombinant IL18 (from cells)
Fournisseur: ProSci Inc.
91-943
PRSI91-943EA 654 EUR InStock
PRSI91-943
Human recombinant IL18 (from cells)
Protéines et Peptides
Interleukin-18 is a secreted protein and it belongs to the IL-1 family. IL-18 is a proinflammatory cytokine and produced by macrophages and other cells. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL-4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein (IL18BP) can specifically interact with this cytokine, and thus negatively regulate its biological activity. After stimulation with IL-18, natural killer (NK) cells and certain T cells release another important cytokine called interferon- gamma (IFN- gamma ) or type II interferon that plays an important role in activating the macrophages or other cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité