Mouse recombinant Ephrin A1
Fournisseur: ProSci Inc.
91-945
PRSI91-945EA 343 EUR InStock
PRSI91-945
Mouse recombinant Ephrin A1
Protéines et Peptides
Ephrin-A1 is a cell membrane protein and contains 1 ephrin RBD (ephrin receptor-binding) domain. EFNA1 belongs to the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNA class ephrin which binds to the EPHA2, EPHA4, EPHA5, EPHA6, and EPHA7 receptors. Two transcript variants that encode different isoforms were identified through sequence analysis.It belongs to the ephrin family and contains 1 ephrin RBD (ephrin receptor-binding) domain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité