Mouse recombinant TWEAK receptor
Fournisseur: ProSci Inc.
91-946
PRSI91-946EA 588 EUR InStock
PRSI91-946
Mouse recombinant TWEAK receptor
Protéines et Peptides
Tumor necrosis factor receptor superfamily member 12A(Tnfrsf12a) is a single-pass type I membrane protein and contains 1 TNFR-Cys repeat. It is weak inducer of apoptosis in some cell types.It promotes angiogenesis and it is the proliferation of endothelial cells. It may modulate cellular adhesion to matrix proteins.TNFR binds specifically to tumor necrosis factor (TNF) and blocks its interaction with cell surface TNF receptors. TNF is a naturally occurring cytokine that is involved in normal inflammatory and immune responses. It plays an important role in the inflammatory processes of rheumatoid arthritis (RA), polyarticular-course juvenile rheumatoid arthritis (JRA), and ankylosing spondylitis and the resulting joint pathology. In addition, TNF plays a role in the inflammatory process of plaque psoriasis. Elevated levels of TNF are found in involved tissues and fluids of patients with RA, psoriatic arthritis, ankylosing spondylitis (AS), and plaque psoriasis. Two distinct receptors for TNF (TNFRs), a 55 kilodalton protein (p55) and a 75 kilodalton protein (p75), exist naturally as monomeric molecules on cell surfaces and in soluble forms. Biological activity of TNF is dependent upon binding to either cell surface TNFR.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité