Mouse recombinant death receptor 6
Fournisseur: ProSci Inc.
91-950
PRSI91-950EA 588 EUR InStock
PRSI91-950
Mouse recombinant death receptor 6
Protéines et Peptides
Tumor necrosis factor receptor superfamily member 21(DR6) is a single-pass type I membrane protein and contains 1 death domain and 4 TNFR-Cys repeats. The protein may activate NF-kappa-B and promote apoptosis and it may activate JNK and be involved in T-cell differentiation.It is required for both normal cell body death and axonal pruning. Trophic-factor deprivation triggers the cleavage of surface APP by beta-secretase to release sAPP-beta which is further cleaved to release an N-terminal fragment of APP (N-APP). N-APP binds TNFRSF21 triggering caspase activation and degeneration of both neuronal cell bodies (via caspase-3) and axons (via caspase-6).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité