Human recombinant CD28 (from cells)
Fournisseur: ProSci Inc.
91-952
PRSI91-952EA 343 EUR InStock
PRSI91-952
Human recombinant CD28 (from cells)
Protéines et Peptides
T-cell-specific surface glycoprotein CD28(CD28) is a single-pass typeI membrane protein which contains one Ig-likeV-type (immunoglobulin-like) domain. It belongs to the immunoglobulin(Ig) superfamily. CD28 is one of the molecules expressed on T cells that provide co-stimulatory signals, which are required for T cell activation.CD28 co-stimulation is necessary for CD4 positive T-cell proliferation and survival, interleukin-2 production, and T-helper type-2 development. Human post-thymic regulatory T cells require CD28 co-stimulation to expand and maintain potent suppressive function in vivo. Apoptosis plays a key role in the age-related decline of CD28 expression and in immunosenescence. CD28 is the receptor for CD80 (B7.1) and CD86 (B7.2). When activated by Toll-like receptor ligands, the CD80 expression is upregulated in antigen presenting cells (APCs). The CD86 expression on antigen presenting cells is constitutive. CD28 is the only B7 receptor constitutively expressed on naive T cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité