Mouse recombinant PSGL-1
Fournisseur: ProSci Inc.
91-961
PRSI91-961EA 588 EUR InStock
PRSI91-961
Mouse recombinant PSGL-1
Protéines et Peptides
P-selectin glycoprotein ligand 1 is the high affinitycounter-receptor for P-selectin on expressed on activated endothelial cells and platelets. As such, it plays a critical role in the tethering of these cells to activated platelets or endothelia expressing P-selectin. As cell adhesion molecules, multiple studies have shown that PSGL-1/ P-selectin interaction is required for the normal recruitment of leukocytes during inflammatory reactions, and also participates in hemostatic responses. PSGL-1 can also bind to other two members of the selectin family, E-selectin (endothelial) and L-selectin (leukocyte), but binds best to P-selectin.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité