Human recombinant IZUMO4 (from cells)
Fournisseur: ProSci Inc.
91-963
PRSI91-963EA 654 EUR InStock
PRSI91-963
Human recombinant IZUMO4 (from cells)
Protéines et Peptides
Izumo sperm-egg fusion protein 4 is a sperm membrane protein which plays a key role in the fusion in the mouse. IZUMO4 has an N-terminal domain with significant homology to the N-terminal domain of Izumo.It belongs to the Izumo family . Izumo 4 is a soluble protein expressed in the testis and in other tissues. Izumo domain possesses the ability to form dimers, whereas the transmembrane domain or the cytoplasmic domain or both of Izumo 1 are required for the formation of multimers of higher order.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité