Human Recombinant NGAL (from Cells)
Fournisseur: ProSci Inc.
91-967
PRSI91-967EA 588 EUR InStock
PRSI91-967
Human Recombinant NGAL (from Cells)
Protéines et Peptides
LCN2 is iron-trafficking protein involved in multiple processes such as apoptosis, innate immunity and renal development. LCN2 binds iron through association with 2,5-dihydroxybenzoic acid (2,5-DHBA), a siderophore that shares structural similarities with bacterial enterobactin, and delivers or removes iron from the cell, depending on the context. LCN2 is involved in apoptosis due to interleukin-3 (IL3) deprivation: iron-loaded form increases intracellular iron concentration without promoting apoptosis, while iron-free form decreases intracellular iron levels, inducing expression of the proapoptotic protein BCL2L11/BIM, resulting in apoptosis. LCN2 is involved in innate immunity, possibly by sequestrating iron, leading to limit bacterial growth.
  • Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité