Human recombinant GDP-L-Fucose Synthase (from E. coli)
Fournisseur: ProSci Inc.
91-969
PRSI91-969EA 789 EUR InStock
PRSI91-969
Human recombinant GDP-L-Fucose Synthase (from E. coli)
Enzymes
GDP-L-Fucose Synthase is a NADP(H)-binding protein. It catalyses the two-step epimerase and the reductase reactions in GDP-D-mannose metabolism, converting GDP-4-keto-6-D-dexoymannose to GDP-L-fucose. GDP-L-Fucose is the substrate of several fucosyltransferase, involving the expression of mamy glycoconjugates, including blood group ABH antigens and development adhesion antigens. Mutations in the TSTA3 gene may cause leukocyte adhesion deficiency type II.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité