Human Recombinant Glutamine Synthetase (from E. coli)
Fournisseur: ProSci Inc.
91-970
PRSI91-970EA 789 EUR InStock
PRSI91-970
Human Recombinant Glutamine Synthetase (from E. coli)
Enzymes
Glutamine Synthetase reglutes intracellular concentration of glutamate. Glutamine Synthetase catalyses the synthesis of glutamine from glutamate and ammonia. Glutamine is an important source of energy and that takes part in cell prolifetation, inhibition of apoptosis, and cell signaling. Glutamine Synthetase is expressed during early fetal stages, and has a role in maintaining body PH by removing ammonia from circulation. Mutations in the GLUL gene are related to congenital glutamine deficiency.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité