Human recombinant PI-TP-alpha (from E. coli)
Fournisseur: ProSci Inc.
91-977
PRSI91-977EA 884 EUR InStock
PRSI91-977
Human recombinant PI-TP-alpha (from E. coli)
Protéines et Peptides
Phosphatidylinositol Transfer Protein alpha Isoform (PITPNA) is found in the cytoplasm and belongs to the PtdIns transfer protein family. PITPNA is a ubiquitous and highly conserved protein in multicellular eukaryotes that catalyzes the exchange of phospholipids between membranes and participates in cellular phospholipid metabolism, signal transduction and vesicular trafficking in vivo. It is expressed in a wide range of tissues and implicated in phospholipase C signaling and in the production of phosphatidylinositol 3, 4, 5-trisphosphate (PIP3) by phosphoinositide-3-kinase.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité