Human recombinant Estrogen Receptor alpha (from E. coli)
Fournisseur: ProSci Inc.
91-978
PRSI91-978EA 852 EUR InStock
PRSI91-978
Human recombinant Estrogen Receptor alpha (from E. coli)
Protéines et Peptides
Estrogen Receptor is a major ligand-activated transcription factor belonging to the nuclear hormone receptor superfamily. Estrogen Receptor is composed of several domains important for hormone binding, DNA binding, and activation of transcription. The protein localizes to the nucleus where it may form a homodimer or a heterodimer with estrogen receptor 2. Estrogen and its receptors are essential for sexual development and reproductive function, but they also play a role in other tissues such as bone. Estrogen receptors are also involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. Alternative splicing results in several transcript variants, which differ in their 5' UTRs and use different promoters.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité