Human recombinant prefoldin subunit 4 (from E. coli)
Fournisseur: ProSci Inc.
91-980
PRSI91-980EA 852 EUR InStock
PRSI91-980
Human recombinant prefoldin subunit 4 (from E. coli)
Protéines et Peptides
Prefoldin Subunit 4 (PFDN4) is a heterohexameric chaperone protein that belongs to the prefoldin subunit beta family. The complex of PFDN4, consisting of two PFD-alpha type and four PFD-beta type subunits, forms a double beta barrel assembly with six protruding coiled-coils. PFDN4 binds and stabilizes newly synthesized polypeptides, thereby allowing them to fold correctly.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité