Human recombinant CLIC4 (from E. coli)
Fournisseur: ProSci Inc.
91-981
PRSI91-981EA 884 EUR InStock
PRSI91-981
Human recombinant CLIC4 (from E. coli)
Protéines et Peptides
Chloride Intracellular Channel Protein 4 (CLIC4) is a 253 amino acid single-pass membrane protein that localizes to both the nucleus and the cytoplasm and contains one GST C-terminal domain. CLIC4 is expressed in various tissues and exhibits an intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells). CLIC4 acts as a monomer which is able to form selective ion channels in target proteins, thus facilitating the transport of chloride and other ions. CLIC4 is believed to have a role in apoptosis and is able to translocate to the nucleus under stress conditions. CLIC4 has alternate cellular functions like a potential role in angiogenesis or in maintaining apical-basolateral membrane polarity during mitosis and cytokinesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité