Human recombinant APBA3 (from E. coli)
Fournisseur: ProSci Inc.
91-983
PRSI91-983EA 852 EUR InStock
PRSI91-983
Human recombinant APBA3 (from E. coli)
Protéines et Peptides
Amyloid beta A4 Precursor Protein-Binding Family A Member 3 (APBA3) is an adapter protein that belongs to the X11 family. APBA3 contains 2 PDZ (DHR) domains and 1 PID domain and interacts with the Alzheimer's disease amyloid precursor protein.. APBA3 is believed to be involved in signal transduction processes. Unlike X11- alpha and - beta which are generally neuronal proteins, APBA3 is widely expressed in all tissues examined with lower levels in brain and testis. It binds to the cytoplasmic domain of amyloid protein (APP) in vivo and may modulate processing of the beta -amyloid precursor protein (APP) and hence formation of beta -APP.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité