Human recombinant SOD1 (from E. coli)
Fournisseur: ProSci Inc.
91-985
PRSI91-985EA 884 EUR InStock
PRSI91-985
Human recombinant SOD1 (from E. coli)
Protéines et Peptides
Superoxide Dismutase [Cu-Zn] (SOD1) is a soluble cytoplasmic and mitochondrial intermembrane space protein that belongs to the Cu-Zn superoxide dismutase family. SOD1 binds copper and zinc ions and is one of three isozymes responsible for destroying free superoxide radicals in the body. SOD1 neutralizes supercharged oxygen molecules, which can damage cells if their levels are not controlled. The enzyme protects the cell against dangerous levels of superoxide. Zinc binding promotes dimerization and stabilizes the native form. Mutations in SOD1 cause a form of familial amyotrophic lateral sclerosis. Defects in SOD1 are the cause of amyotrophic lateral sclerosis type 1 (ALS1) which is a familial form of amyotrophic lateral sclerosis, a neurodegenerative disorder affecting upper and lower motor neurons and resulting in fatal paralysis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité