Human recombinant WW domain-binding 2 (from E. coli)
Fournisseur: ProSci Inc.
91-988
PRSI91-988EA 852 EUR InStock
PRSI91-988
Human recombinant WW domain-binding 2 (from E. coli)
Protéines et Peptides
WW Domain-Binding Protein 2 (WBP2) is a ubiquitous protein that contains one GRAM domain. The WW domain is composed of 38 to 40 semi-conserved AA shared by proteins of diverse functions including structural, regulatory, and signaling proteins. The domain is participated in mediating protein-protein interactions. WBP2 binds to the WW domain of YAP1, WWP1 and WWP2. The WW-binding 1 motif of WBP2 mediates interaction with NEDD4. The function of this protein WBP2 has not been determined. Some researches demonstrate that WBP-2 also interacts with the thyroid-specific transcription factor Pax8.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité