Human recombinant partner of Y14 and mago (from E. coli)
Fournisseur: ProSci Inc.
91-994
PRSI91-994EA 789 EUR InStock
PRSI91-994
Human recombinant partner of Y14 and mago (from E. coli)
Protéines et Peptides
Partner of Y14 and Mago (WIBG) is a key regulator of the Exon Junction Complex (EJC). EJC is a multiprotein complex that associates immediately upstream of the exon-exon junction on mRNAs, is a positional landmarker for the intron exon structure of genes, and directs post-transcriptional processes in the cytoplasm, for instance mRNA export, nonsense-mediated mRNA decay or translation. WIBG is a cytoplasmic RNA-binding protein, it can be excluded from nucleus by Crm1. WIBG as a cooperateing partner of Mago-14, relates with Mago-14 by its N-terminal domain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité