Human recombinant Autophagy 3 (from E. coli)
Fournisseur: ProSci Inc.
91-995
PRSI91-995EA 343 EUR InStock
PRSI91-995
Human recombinant Autophagy 3 (from E. coli)
Protéines et Peptides
Ubiquitin-Like-Conjugating Enzyme ATG3 (ATG3) is widely expressed and has highly levels in heart, skeletal muscle, kidney, liver and placenta. ATG3 as a E2-like enzyme, involves in autophagy and mitochondrial homeostasis. ATG3 catalyzes the conjugation of ATG8-like proteins to PE which is essential for autophagy. As an autocatalytic E2-like enzyme, ATG3 also can catalyzes the conjugation of ATG12 to itself which palys a role in mitochondrial homeostasis but not in autophagy.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His, N-T7 tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité