Human recombinant Autophagy 10 (from E. coli)
Fournisseur: ProSci Inc.
91-998
PRSI91-998EA 884 EUR InStock
PRSI91-998
Human recombinant Autophagy 10 (from E. coli)
Protéines et Peptides
Ubiquitin-Like-Conjugating Enzyme ATG10 (ATG10) is a ubiquitous 28kDa member of the ATG10 family protein. ATG10 acts as an E2-like enzyme, catalyzes the transfer of ATG12 to ATG5 during in the initial stages of autophagesome formation. The heterodimer of ATG5 and ATG12 subsequntly associates non-covalently with an ATG16 multimer to generate an antophagosome. ATG10 plays a role in the conjugation of ATG12 to ATG5 by interaction with MAP1LC3A. In addition, ATG10 can diretly interact with ATG5 or ATG7.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His, N-T7 tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité