Human Recombinant Glycine N-Methyltransferase (from E. coli)
Fournisseur: ProSci Inc.
91-999
PRSI91-999EA 789 EUR InStock
PRSI91-999
Human Recombinant Glycine N-Methyltransferase (from E. coli)
Enzymes
Glycine N-Methyltransferase (GNMT) is a tetrameric cytosolic protein. GNMT catalyses the synthesis of N-methylglycine from glycine using S-adenosylmethionine (AdoMet) as the methyl donor. It can affects DNA methylation by regulating the ratio of S-adenosylmethionine to S-adenosylhomocystine, playing an important role in maintaining normal AdoMet levels. GNMT is highly expressed in liver. As a major folate-binding protein, GNMT takes part in the detoxification pathway. Defects in GNMT are the cause of hypermethioninemia. the patients with this deficiency are mild hepatomegaly and chronic elevation of serum transaminases.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité