Human recombinant Galactokinase (from E. coli)
Fournisseur: ProSci Inc.
92-004
PRSI92-004EA 789 EUR InStock
PRSI92-004
Human recombinant Galactokinase (from E. coli)
Enzymes
Galactokinase belongs to the GHMP kinase family and GalK subfamily. Galactokinase is more important to the galactose metabolism pathway. It modifies galactose to create a similar molecule called Galactose-1-Phosphate.Thus, a series of additional steps converts Galactose-1-Phosphate to another simple sugar Glucose, which is the main energy source for most cells. Galactokinase deficiency results in Galactosemia II, an autosomal recessive deficiency characterised by congenital cataracts during infancy and presenile cataracts in the adult population. The cataracts are secondary to accumulation of Galactitol in the lenses.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité