Human Recombinant GSTZ1 (from E. coli)
Fournisseur: ProSci Inc.
92-007
PRSI92-007EA 884 EUR InStock
PRSI92-007
Human Recombinant GSTZ1 (from E. coli)
Protéines et Peptides
Maleylacetoacetate Isomerase (MAAI) belongs to the Glutathione S-Transferase super-family. MAAI encodes multifunctional enzymes in the detoxification of electrophilic molecules by conjugation with glutathione, for example, mutagens, carcinogens and several therapeutic drugs. MAAI is a bifunctional protein with low glutathione peroxidase activity with T-butyl and cumene hydroperoxides. MAAI can catalyze the glutathione dependent oxygenation of dichloroacetic acid to glyoxylic acid. But it has minimal glutathione-conjugating activity with 7-chloro-4-nitrobenz-2-oxa-1, ethacrynic acid 3-diazole and maleylacetoacetate isomerase activity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité