Human recombinant zinc finger 75A (fromE. coli)
Fournisseur: ProSci Inc.
92-017
PRSI92-017EA 761 EUR InStock
PRSI92-017
Human recombinant zinc finger 75A (fromE. coli)
Protéines et Peptides
Zinc Finger Protein 75A (ZNF75A) is a member of krueppel C2H2-type zinc-finger protein family. The human ZNF75 gene is located on Xq26, which has only limited homology (less than 65%) to other ZF genes in the databases. One of these, ZNF75B is a pseudogene mapped to chromosome 12q13. The other two, ZNF75A and ZNF75C, maintain an ORF in the sequenced region, and at least the latter is expressed in the U937 cell line. ZNF75A contains five C2H2-type zinc fingers and one KRAB domain. ZNF75A is a nucleus protein, may involves in transcriptional regulation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité