Human recombinant Selenocysteine lyase (from E. coli)
Fournisseur: ProSci Inc.
92-018
PRSI92-018EA 789 EUR InStock
PRSI92-018
Human recombinant Selenocysteine lyase (from E. coli)
Enzymes
Selenocysteine Lyase belongs to the class-V pyridoxal-phosphate-dependent aminotransferase family. Selenocysteine Lyase exists as a homodimer in the cytosol. In the brain, Selenocysteine Lyase is as an enzyme that putatively salvages Sec and recycles the selenium for selenoprotein translation. Selenocysteine Lyase catalyses the decomposition of L-selenocysteine to L-alanine and elemental selenium. Selenocysteine Lyase can be up-regulated In acute glomerulonephritis, it can also be regulated by JUN/AP-1.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité