Human recombinant ZBED1 (from E. coli)
Fournisseur: ProSci Inc.
92-021
PRSI92-021EA 852 EUR InStock
PRSI92-021
Human recombinant ZBED1 (from E. coli)
Protéines et Peptides
Zinc Finger BED Domain-Containing Protein 1 (ZBED1) contains one BED-type zinc finger and is found in the cell nucleus. ZBED1 is widely expressed, highly in heart, skeletal muscle, spleen and placenta. The expression of ZBED1 is usually linked to the cell cycle. During the G1/S phase, the expression is increasing. During the S/G2 phage, the expression reaches to the highest, and then decreasing. ZBED1 exists in homodimer forms, which can bind to 5’-TGTCCG[CT]GA[CT]A-3’ DNA elements, that can be found in the promoter regions of a number of gene related to cell proliferation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité