Human recombinant Testin (from E. coli)
Fournisseur: ProSci Inc.
92-024
PRSI92-024EA 654 EUR InStock
PRSI92-024
Human recombinant Testin (from E. coli)
Protéines et Peptides
Testin belongs to the prickle/espinas/testin family. Testin contains three LIM zinc-binding domains and one PET domain, TES as a scaffold protein that may have a role in cell adhesion, cell spreading and in the reorganization of the actin cytoskeleton. In addition, TES can also as a tumor suppressor, inhibits tumor cell growth, regulate the cell proliferation.TES interacts with many cytoskeletal proteins, such as Zyxin, Mena, Actin, Talin and VASP. The ability of TES to associate with alpha-actin, Zyxin and paxillin is dependent on the conformational state of the molecule.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité