Human Recombinant High Mobility Group B1 (from E. coli)
Fournisseur: ProSci Inc.
92-029
PRSI92-029EA 654 EUR InStock
PRSI92-029
Human Recombinant High Mobility Group B1 (from E. coli)
Protéines et Peptides
High mobility group protein B1 is a member of the HMGB family consisting of three members, HMGB1, HMGB2 and HMGB3.It Contains 2 HMG box DNA-binding domains entitled box A and box B and It is a highly negative-charged C terminus. As a nuclear protein, HMGB1 stabilizes nucleosomes and allows bending of DNA that facilitates gene transcription which is essential for individual survival. Meanwhile, it is revealed that HMGB1 can also act as a cytokine extracellularlly and regulates monocyte, T cell, dendritic cell activities in inflammatory responses.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité