Human recombinant BCAS2 (from E. coli)
Fournisseur: ProSci Inc.
92-031
PRSI92-031EA 852 EUR InStock
PRSI92-031
Human recombinant BCAS2 (from E. coli)
Protéines et Peptides
Breast Carcinoma-Amplified Sequence 2 (BCAS2) is a member of the SPF27 family. BCAS2 is a nuclear protein and widely expressed in many rtissues. BCAS2 is identified as being overexpressed in various breast cancer cell lines. BCAS2 is a component of the spliceosome, taking part in the removal of introns from mRNA precursors. BCAS2 interacts with estrogen receptor alpha and beta, thyroid hormone receptor beta, peroxisome proliferator-activated receptor gamma. BCAS2 functions as an ER co-activator and is capable of enhancing ER-mediated transcription.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His, N-T7 tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité