Human recombinant zinc finger 70 (fromE. coli)
Fournisseur: ProSci Inc.
92-033
PRSI92-033EA 852 EUR InStock
PRSI92-033
Human recombinant zinc finger 70 (fromE. coli)
Protéines et Peptides
Zinc Finger Protein 70 (ZNF70) is a member of the krueppel C2H2-type zinc-finger protein family. ZFN70 is localized to the cell nucleus and contains eleven C2H2-type zinc fingers. ZFN70 has sequence-specific DNA binding transcription factor activity, and it may be involved in transcriptional regulation. In addition, ZFN70 has the DNA-binding and metal-binding activity, which can bind zinc ion.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité