Human recombinant Thioredoxin (from E. coli)
Fournisseur: ProSci Inc.
92-045
PRSI92-045EA 343 EUR InStock
PRSI92-045
Human recombinant Thioredoxin (from E. coli)
Protéines et Peptides
Thioredoxin (TXN) is a member of the Thioredoxin family. Thioredoxin exists as a disulfide-linked homodimer and contains one Thioredoxin domain. Thioredoxin is up-regulated by ionizing radiation. Thioredoxin participates in various redox reactions through the reversible oxidation of its active center dithiol to a disulfide and catalyzes dithiol-disulfide exchange reactions. Thioredoxin also plays a role in the reversible S-nitrosylation of cysteine residues in target proteins, and thereby contributes to the response to intracellular nitric oxide.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité