Human recombinant Catalase (from E. coli)
Fournisseur: ProSci Inc.
92-046
PRSI92-046EA 789 EUR InStock
PRSI92-046
Human recombinant Catalase (from E. coli)
Enzymes
Catalase (CAT) is a member of the catalase family. It exists as a homotetramer that occurs in almost all aerobically respiring organisms and serves to protect cells from the toxic effects of hydrogen peroxide. Catalase is localised in the peroxisome. Catalase promotes growth of cells including T-cells, B-cells, myeloid leukemia cells, melanoma cells, mastocytoma cells, and normal and transformed fibroblast cells. Defects in CAT are the cause of acatalasemia which is characterised by absence of catalase activity in red cells and is associated with ulcerating oral lesions.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité