Human recombinant MAP1LC3A (from E. coli)
Fournisseur: ProSci Inc.
92-048
PRSI92-048EA 584 EUR InStock
PRSI92-048
Human recombinant MAP1LC3A (from E. coli)
Protéines et Peptides
Microtubule-Associated Proteins 1A/1B Light Chain 3A (MAP1LC3A) belongs to the MAP1 LC3 family. MAP1LC3A is found most abundantly in the heart, brain, liver, skeletal muscle, and testis. But it is absent in the thymus and peripheral blood leukocytes. MAP1LC3A is thought to take part in the formation of autophagosomal vacuoles and is one of the light chain subunits that functions together with both MAP1A and/or MAP1B. In addition, MAP1A has an important part in neuronal development and in maintaining the balance between neuronal plasticity and rigidity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité