Human recombinant alpha-Parvin (from E. coli)
Fournisseur: ProSci Inc.
92-050
PRSI92-050EA 884 EUR InStock
PRSI92-050
Human recombinant alpha-Parvin (from E. coli)
Protéines et Peptides
Alpha-Parvin (PARVA) is a member of the Parvin family. PARVA contains two CH (calponin-homology) domains. PARVA is widely expressed, with highest levels in heart, skeletal muscle, kidney and liver. PARVA interacts with integrin-linked protein kinase and probably with actin and the LD1 and LD4 motifs of PXN. PARVA may play a role in the regulation of cell adhesion and cytoskeleton organization. PARVA is also involved in ciliogenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité