Human Recombinant DFF45 (from E. coli)
Fournisseur: ProSci Inc.
92-051
PRSI92-051EA 588 EUR InStock
PRSI92-051
Human Recombinant DFF45 (from E. coli)
Protéines et Peptides
DNA Fragmentation Factor Subunit Alpha (DFFA). DFFA exists as a heterodimer (DFF) with DFFB. DFF is activated once DFFA is cleaved by Caspase-3. The cleaved fragments of DFFA detach from DFFB (the active component of DFF), which in turn triggers DNA fragmentation as well as chromatin condensation during apoptosis. A reduced level of DFFA detected in ovarian endometriosis may be a part of an apoptosis-resistant mechanism enhancing the disease progression.
  • Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité