Human recombinant XPNPEP3 (from E. coli)
Fournisseur: ProSci Inc.
92-052
PRSI92-052EA 789 EUR InStock
PRSI92-052
Human recombinant XPNPEP3 (from E. coli)
Protéines et Peptides
Probable Xaa-Pro Aminopeptidase 3 (XPNPEP3) is a member of the peptidase M24B family. XPNPEP3 has two isoforms and both are widely expressed. XPNPEP3 is localized in the Mitochondrion. XPNPEP3 catalyzes the release of any N-terminal amino acid, including proline, that is linked to proline, even from a dipeptide or tripeptide. Defects in XPNPEP3 are the cause of nephronophthisis-like nephropathy type 1 which is a disorder with features of nephronophthisis, a cystic kidney disease leading to end-stage renal failure.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N, C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité