Human recombinant PPIase D (from E. coli)
Fournisseur: ProSci Inc.
92-055
PRSI92-055EA 584 EUR InStock
PRSI92-055
Human recombinant PPIase D (from E. coli)
Enzymes
Peptidyl-Prolyl Cis-Trans Isomerase D (PPID) belongs to the cyclophilin-type PPIase family and PPIase D subfamily. PPID is widely expressed and it contains one PPIase cyclophilin-type domain and three TPR repeats. PPID catalyses the cis-trans isomerisation of proline imidic peptide bonds in oligopeptides and accelerates the folding of proteins. PPID can bind to the immunosuppressant cyclosporine A and is known that its overexpression suppresses the apoptosis in cancer cells.
Fusion-Tag: N, C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité