Human recombinant IGF2 mRNA-Binding 2 (from E. coli)
Fournisseur: ProSci Inc.
92-057
PRSI92-057EA 852 EUR InStock
PRSI92-057
Human recombinant IGF2 mRNA-Binding 2 (from E. coli)
Protéines et Peptides
Insulin-Like Growth Factor 2 mRNA-Binding Protein 2 (IGFBP2) belongs to the RRM IMP/VICKZ family. IGFBP2 is a cytoplasmic protein and contains four KH domains and two RRM (RNA recognition motif) domains. IGF2BP2 binds to the 5'-UTR of the Insulin-Like Growth Factor 2 (IGF2) mRNA. This binding is isoform-specific. IGF2BP2 may regulate translation of target mRNAs. Genetic variation at the IGF2BP2 gene has been associated with type 2 diabetes (T2D) by genome-wide association studies and by replication analyses.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His, N-T7 tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité