Human recombinant Annexin A10 (from E. coli)
Fournisseur: ProSci Inc.
92-062
PRSI92-062EA 852 EUR InStock
PRSI92-062
Human recombinant Annexin A10 (from E. coli)
Protéines et Peptides
Annexin A10 (ANXA10) contains four Annexin repeats and is a member of the Annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. It is reported that ANXA10 may be a clinical relevant marker for predicting outcome in both early and advanced stages of bladder cancer.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité