Human recombinant FGF1 Intracellular-Binding (from E. coli)
Fournisseur: ProSci Inc.
92-063
PRSI92-063EA 884 EUR InStock
PRSI92-063
Human recombinant FGF1 Intracellular-Binding (from E. coli)
Protéines et Peptides
Acidic Fibroblast Growth Factor Intracellular-Binding Protein (FIBP) is highly expressed in the heart, skeletal muscle, and pancreas. Acidic Fibroblast Growth Factor is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. FIBP is an intracellular protein that binds selectively to acidic fibroblast growth factor (aFGF). It is postulated that FIBP may be involved in the mitogenic action of aFGF.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité