Human recombinant PADI4 (from E. coli)
Fournisseur: ProSci Inc.
92-065
PRSI92-065EA 789 EUR InStock
PRSI92-065
Human recombinant PADI4 (from E. coli)
Protéines et Peptides
Protein-Arginine Deiminase Type-4 (PADI4) belongs to the Protein Arginine Deiminase family. PADI4 is expressed in eosinophils and neutrophils, but it is not expressed in peripheral monocytes or lymphocytes. PADI4 catalyzes the citrullination/deimination of arginine residues of proteins. In addition, PADI4 may play an important role in tumourigenesis. Genetic variations in PADI4 are a cause of susceptibility to rheumatoid arthritis (RA), which is a systemic inflammatory disease with autoimmune features and a complex genetic component.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité