Human recombinant BDH1 (from E. coli)
Fournisseur: ProSci Inc.
92-066
PRSI92-066EA 884 EUR InStock
PRSI92-066
Human recombinant BDH1 (from E. coli)
Protéines et Peptides
D-Beta-Hydroxybutyrate Dehydrogenase Mitochondrial (BDH) is a member of the short-chain dehydrogenases/reductases (SDR) family. BDH is localized in the mitochondrion matrix. BDH forms a homotetrameric lipid-requiring enzyme of the mitochondrial membrane and has a specific necessity for phosphatidylcholine for optimal enzymatic activity. BDH catalyzes the interconversion of acetoacetate and (R)-3-hydroxybutyrate, the 2 main ketone bodies formed during fatty acid catabolism.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité