Mouse recombinant TNF alpha (from E. coli)
Fournisseur: ProSci Inc.
92-067
PRSI92-067EA 727 EUR InStock
PRSI92-067
Mouse recombinant TNF alpha (from E. coli)
Protéines et Peptides
Tumor Necrosis Factor (TNF) is a member of the Tumor Necrosis Factor family. TNF exists as a homotrimer and interacts with SPPL2B. TNF is mainly secreted by macrophages and can induce cell death of certain tumor cell lines. TNF is a key cytokine in the development of several inflammatory disorders. It contributes to the development of type 2 diabetes throught its effects on insulin resistance and fatty acid metabolism.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité