Human recombinant serum amyloid A4 (from E. coli)
Fournisseur: ProSci Inc.
92-072
PRSI92-072EA 654 EUR InStock
PRSI92-072
Human recombinant serum amyloid A4 (from E. coli)
Protéines et Peptides
Serum Amyloid A-4 Protein (SAA4) is a member of the SAA family. SAA proteins are a family of apolipoproteins associated with high-density lipoprotein (HDL) in plasma. SAA4 is constitutively expressed only in humans and mice. Its physiological function is unknown. SAA4 functions as a major acute phase reactant. SAA4 mRNA and protein occurrence in macrophage derived foam cells of coronary and carotid arteries implied a specific role of human SAA4 during inflammation including atherosclerosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité