Human recombinant TRIM (from E. coli)
Fournisseur: ProSci Inc.
92-074
PRSI92-074EA 884 EUR InStock
PRSI92-074
Human recombinant TRIM (from E. coli)
Protéines et Peptides
T-Cell Receptor-Associated Transmembrane Adapter 1 (TRAT1) is a single-pass type III membrane protein. TRAT1 exists as a disulfide-linked homodimer and is strongly expressed in the thymus, and to a lesser extent in the spleen, lymph node, and peripheral blood lymphocytes. TRAT1 is phosphorylated on tyrosines by LCK or FYN upon TCR activation. Its function is to stabilizes the TCR (T-cell antigen receptor)/CD3 complex at the surface of T-cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité