Human recombinant Syntaxin 6 (from E. coli)
Fournisseur: ProSci Inc.
92-076
PRSI92-076EA 852 EUR InStock
PRSI92-076
Human recombinant Syntaxin 6 (from E. coli)
Protéines et Peptides
Syntaxin-6 (STX6) is a single-pass type IV membrane protein, which belongs to the Syntaxin family. STX6 is mainly localized in the plasma membrane. STX6 contains one t-SNARE coiled-coil homology domain and involved in intracellular vesicle trafficking. When STX6 function is inhibited, internalization through caveolae is dramaticaliy reduced, whereas other endocytic mechanisms are unaffected. It is reported that STX6 is necessary for proper expression of focal adhesion kinase and integrin alpha5 adhesion receptor.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité